missing translation for 'onlineSavingsMsg'
Learn More

Eganelisib, MedChemExpress

Product Code. 16425867
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425867

Brand: Medchem Express HY100716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eganelisib (IPI549) is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. Eganelisib shows >100-fold selectivity over other lipid and protein kinases.

Eganelisib (IPI549) is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. Eganelisib shows >100-fold selectivity over other lipid and protein kinases.

Specifications

Chemical Name or Material Eganelisib
Physical Form Powder
CAS 1693758-51-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C30H24N8O2
For Use With (Application) Cancer-Kinase/protease
Synonym IPI-549
Solubility Information DMSO : 15 mg/mL (28.38 mM; Need ultrasonic and warming)
SMILES O=C(C1=C2N=CC=CN2N=C1N)N[C@H](C3=CC4=C(C(N3C5=CC=CC=C5)=O)C(C#CC6=CN(C)N=C6)=CC=C4)C
Molecular Weight (g/mol) 528.56
Formula Weight 528.56
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.