missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EFHC1 Polyclonal antibody specifically detects EFHC1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | EFHC1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | dJ304B14.2, EF-hand domain (C-terminal) containing 1, EF-hand domain-containing protein 1, EJA1, EJM1, FLJ10466, FLJ37290, JAE, Myoclonin-1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 391-640 of human EFHC1 (NP_060570.2).,, Sequence:, EDSAQNCFALIPKAPKKDVIKMLVNDNKVLRYLAVLESPIPEDKDRRFVFSYFLATDMISIFEPPVRNSGIIGGKYLGRTKVVKPYSTVDNPVYYGPSDFFIGAVIEVFGHRFIILDTDEYVLKYMESNAAQYSPEALASIQNHVRKREAPAPEAESKQTEKDPGVQELEALIDTIQKQLKDHSCKDNIREAFQIYDKEASGYVDRDMFFKICESLNVPVDDSLVKELIRMCSHGEGKINYYNFVRAFSN |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?