Learn More
Ansell EDGE™ 48-969 Protective Gloves
Product Code. 18816165 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
Unit Size:
144 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
18816165 10 144 pairs
18876155 6 144 pairs
18886155 7 144 pairs
18896155 8 144 pairs
18806165 9 144 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 18816165 Supplier Ansell Supplier No. 4896910
Please to purchase this item. Need a web account? Register with us today!
Economical solution for medium duty applications requiring very high cut resistance, grip and oil-repellence performance in oily environment.
  • Heightened grip: Featuring a fully NBR-coated seamless liner with a rough palm, EDGE™ 48-969 protective gloves offer a highly secure grip in oily environments, while protecting skin from injuries and oil contamination
  • Optimized visibility: these bright yellow cut-resistant gloves ensure that hands remain highly visible, promoting safety even in low-light or dark conditions
  • Enhanced defenses: EDGE™ 48-969 offers a high level of abrasion resistance and very high cut protection. In addition, the glove provides EN 407 level 1 contact heat protection, making it ideal for workers handling hot equipment, parts and objects.
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Nitrile
Color Yellow
Cut Resistance Rating Level F
Hand Specific
Latex Free Yes
Material HPPE, Nitrile, Spandex, Nylon, Steel Fiber
Product Type Industrial Gloves
Size 10