missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA-A1/Ectodysplasin A1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81908AF700
This item is not returnable.
View return policy
Description
EDA-A1/Ectodysplasin A1 Polyclonal antibody specifically detects EDA-A1/Ectodysplasin A1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| EDA-A1/Ectodysplasin A1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| EDA-A1 antibody was raised against an 19 amino acid peptide near the center of human EDA-A1. The immunogen is located within amino acids 130 - 180 of EDA-A1. Amino Acid Squence: PDEKPYSEEESRRVRRNKR | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 1896 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction