Learn More
Ansell™ Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves
Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
Description
- Specially treated to reduce the risk of allergic reactions.
- Thickness of 0.35 mm, provides outstanding sensitivity.
- Excellent price/quality ratio.
Specifications
Specifications
| Back Material | Rubber Latex |
| Cuff Material | Natural Rubber Latex |
| Applications | Industrial |
| Coating Material | Natural Rubber Latex |
| Color | Yellow |
| Latex Free | No |
| Material | Rubber |
| Product Type | Extra Thin Gloves |
| Size | 7 |
| Thickness (Metric) | 0.35 mm |
| Show More |