Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 45 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE15549
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 15270464
  • 184.00 € / Each
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Description
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Specifications
8 x 6
Aluminum
45 mm
Microtiterplates (48)
Upright Freezers