Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 45 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE15490
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 12428240
  • 153.00 € / Each
Estimated Shipment: 18-09-2024
to see stock.
Add to basket
Description
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Specifications
8 x 5
Aluminum
450 x 135 x 372 mm
Microtiterplates (40)
Upright Freezers
45 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers