Learn More
Elma™ EC 40 Cleaner
Click to view available options
Volume (Metric):
1 L
Container Type:
Bottle
Product Type:
EC 25 Cleaner
Unit Size:
1 L
Description
- Removes fast and efficiently dental cements from bridges, crowns, dental and medical instruments made from: stainless-steel, ceramics, plastics, glass and rubber.
- Removes dental cements including phosphate cements, metal oxides, lime and fluxing agents.
- Not suitable for light metals (e.g. AI, Mg, Zn and corresponding alloys).
Specifications
Specifications
| Product Type | EC 25 Cleaner |
| Certifications/Compliance | CE |
| Container Type | Bottle |
| For Use With (Application) | For the thorough ultrasonic cleaning of impression trays and dental instruments made from Stainless Steel, Glass, Ceramics and Plastics. |
| Volume (Metric) | 1 L |