Learn More
Elma™ EC 40 Cleaner
Product Code. 10475632
Change view
Click to view available options
Volume (Metric):
1 L
Container Type:
Bottle
Product Type:
EC 25 Cleaner
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Container Type Product Type unitSize
10475632 1 L Bottle EC 25 Cleaner 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10475632 Supplier Elma™ Supplier No. 5813700000
Please to purchase this item. Need a web account? Register with us today!
Providing powerful cleaning power for bridges, crowns, dental and medical instruments made from: stainless-steel, ceramics, plastics, glass and rubber.
  • Removes fast and efficiently dental cements from bridges, crowns, dental and medical instruments made from: stainless-steel, ceramics, plastics, glass and rubber.
  • Removes dental cements including phosphate cements, metal oxides, lime and fluxing agents.
  • Not suitable for light metals (e.g. AI, Mg, Zn and corresponding alloys).
Specifications
Product Type EC 25 Cleaner
Certifications/Compliance CE
Container Type Bottle
For Use With (Application) For the thorough ultrasonic cleaning of impression trays and dental instruments made from Stainless Steel, Glass, Ceramics and Plastics.
Volume (Metric) 1 L