missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EB3 Antibody (KT36), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-23294
This item is not returnable.
View return policy
Description
EB3 Monoclonal antibody specifically detects EB3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| EB3 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| KT36 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| EB1 protein family member 3, EB3EBF3-S, End-binding protein 3, microtubule-associated protein RP/EB family member 3, microtubule-associated protein, RP/EB family, member 3, RP3EBF3APC binding protein | |
| Clone KT36 was generated from a GST fusion protein containing human EB3. This sequence is highly conserved in rat and mouse EB3. [UniProt# Q9UPY8] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, GPCR, Signal Transduction | |
| 22924 | |
| Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction