missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Mouse Plasmacytoid DC Isolation Kit

Product Code. 17117961
Encompass
Change view
Click to view available options
:
2 x 109 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17117961 2 x 109 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17117961 Supplier STEMCELL Technologies Supplier No. 19764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of untouched mouse plasmacytoid dendritic cells (pDC)

  • The EasySep™ Mouse Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from single-cell suspensions of splenocytes or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-pDCs
  • Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran
  • These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube

Specifications

Cell Type Dendritic Cells
For Use With (Application) Cell Isolation
Quantity 2 x 109 cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.