Learn More
STEMCELL Technologies EasySep™ Mouse PE Positive Selection Kit II
Description
- The EasySep™ Mouse PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies
- Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This kit replaces the EasySep™ Mouse PE Positive Selection Kit (Catalog #18554)
Specifications
Specifications
| Cell Type | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 5 x 109 cells |