Learn More
STEMCELL Technologies EasySep™ Mouse PE Positive Selection Kit II
Product Code. 17197741
Encompass
Change view
Click to view available options
Quantity:
1 x 109 cells
5 x 109 cells
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17197741 5 x 109 cells 1 set
17157731 1 x 109 cells 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17197741 Supplier STEMCELL Technologies Supplier No. 17696
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Immunomagnetic positive selection of mouse cells labeled with PE-conjugated antibodies
  • The EasySep™ Mouse PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies
  • Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • This kit replaces the EasySep™ Mouse PE Positive Selection Kit (Catalog #18554)
Specifications
Cell Type B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells
For Use With (Application) Cell Isolation
Quantity 5 x 109 cells