Learn More
STEMCELL Technologies EasySep™ Mouse Monocyte Isolation Kit
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- The EasySep™ Mouse Monocyte Isolation Kit targets non-monocyte cells by labeling unwanted cells with antibodies and magnetic particles, and separates cells without columns using an EasySep™ magnet
- Desired cells are simply poured off into a new tube
- Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays
- This product replaces the EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) for even faster cell isolations
Specifications
Specifications
| Cell Type | Monocytes |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |