missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Mouse Memory CD4+ T Cell Isolation Kit
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- The EasySep™ Mouse Memory CD4+ T Cell Isolation Kit is designed to isolate pan-memory CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies directed against non-memory CD4+ T cells (CD8, CD11b, CD11c, CD19, CD24, CD45R, CD45RB, CD49b, CD105 and TCRγ/δ) and streptavidin-coated magnetic particles (RapidSpheres™ )
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+ |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction