missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human TCR Alpha/Beta Depletion Kit
Click to view available options
Quantity:
1 Kit
missing translation for 'unitSize'
1 set
Description
- The EasySep™ Human TCR Alpha/Beta Depletion Kit is designed to deplete human T cell receptor alpha/beta+ (TCRab+) cells from leukapheresis samples or expanded TCRa/b knockout cell cultures
- TCRab+ cells are targeted with antibody complexes and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Labeled cells remain in the tube while unlabeled cells are poured off
Specifications
Specifications
| Cell Type | T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction