missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Progenitor Cell Enrichment Kit II
Description
- The EasySep™ Human Progenitor Cell Enrichment Kit II is designed to isolate hematopoietic progenitor cells from fresh cord blood and other cell preparations by negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?