missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Cord Blood CD34 Positive Selection Kit III
Description
- Isolate highly purified CD34+ cells from fresh whole umbilical cord blood (CB) using a two-step procedure
- Hematopoietic progenitor cells are pre-enriched using the RosetteSep™ Human Progenitor Cell Basic Pre-Enrichment Cocktail (15226C), with antibodies to remove cells expressing T, B and myeloid cell surface markers
- CD34+ cells are then selected using the EasySep™ Human CD34 Positive Selection Cocktail (18096C)
- f isolating CD34+ cells from whole CB in which platelet contamination is expected the EasySep™ Human Cord Blood CD34 Positive Selection Kit II (Catalog #17896) is recommended
- If isolating CD34+ cells from fresh blood or buffy coat, the Complete Kit for Human Whole Blood CD34+ Cells (Catalog #15086) is recommended
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1000 mL cord blood |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?