missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human Cord Blood CD34 Positive Selection Kit II

Product Code. 17177811
Encompass
Click to view available options
Quantity:
1000 mL Cord Blood
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17177811

Brand: STEMCELL Technologies 17896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Complete kit for CD34+ cells from cord blood

  • Isolate highly purified CD34+ cells from fresh whole umbilical cord blood using a simple, two-step procedure
  • Hematopoietic progenitor cells are pre-enriched using the RosetteSep™ Human Cord Blood CD34 Pre-Enrichment Cocktail (15896C), through the use of antibodies to remove cells expressing T cell, B cell, myeloid cell and platelet surface markers
  • CD34+ cells are then selected using the EasySep™ Human CD34 Positive Selection Cocktail (18096C), which contains an antibody recognizing CD34
  • If isolating CD34+ cells from fresh blood or buffy coat the Complete Kit for Human Whole Blood CD34+ Cells (Catalog #15086) is recommended
  • If isolating CD34+ cells from other samples, including fresh or previously frozen mobilized peripheral blood or bone marrow mononuclear cells, or from previously frozen cord blood mononuclear cells, we recommend using the EasySep™ Human CD34 Positive Selection Kit (Catalog #17856)

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Isolation
Quantity 1000 mL Cord Blood
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.