missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human CD14 Positive Selection Kit II

Product Code. 15947155
Encompass
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15947155

Brand: STEMCELL Technologies 17858

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic positive selection of human CD14+ cells (e.g. monocytes)

  • The EasySep™ Human CD14 Positive Selection Kit II is designed to isolate highly purified CD14+ cells from fresh or previously frozen human peripheral blood mononuclear cells (PBMCs)
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD14 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD14 antigen is strongly expressed on monocytes and macrophages and weakly on granulocytes
  • It is also expressed on most tissue macrophages
  • This product replaces the EasySep™ Human CD14 Positive Selection Kit (Catalog #18058), for even faster cell isolations and is compatible with all EasySep™ magnets

Specifications

Cell Type Monocytes, Myeloid Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.