missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human B Cell Enrichment Kit

Product Code. 15551575
Encompass
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15551575

Brand: STEMCELL Technologies 19054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of untouched human B cells

  • The EasySep™ Human B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and dextran-coated magnetic particles
  • The labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube

Specifications

Cell Type B Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 Cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.