Learn More
STEMCELL Technologies EasySep™ Direct Human Monocyte Isolation Kit
Click to view available options
Quantity:
100 mL whole blood
Unit Size:
Each
Description
- The EasySep™ Direct Human Monocyte Isolation Kit isolates functional, highly purified CD14+ monocytes directly from human whole blood by immunomagnetic negative selection
- No lysis, density gradient centrifugation or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture and other downstream applications
Specifications
Specifications
| Cell Type | Monocytes, Myeloid Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 100 mL whole blood |