missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Direct Human B-CLL Cell Isolation Kit
Click to view available options
:
100 mL whole blood
Description
- This kit isolates untouched and highly purified B-CLL cells directly from human whole blood by fully immunomagnetic negative selection
- The depletion cocktail in this kit does not contain antibodies against CD43, which can be expressed on malignant cells in chronic lymphocytic leukemia (CLL) samples
- This cell isolation kit targets non-B cells for removal based on specific surface antigens
- Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet
- Desired cells are simply collected into a new tube and can immediately be used in downstream applications such as nucleic acid extraction, flow cytometry or culture
Specifications
Specifications
| Cell Type | B Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 100 mL whole blood |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction