missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Direct HLA B Cell Isolation Kit

Product Code. 17159951
Encompass
Click to view available options
Quantity:
100 mL whole blood
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17159951

Brand: STEMCELL Technologies 89684

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of untouched human B cells for HLA analysis - 100 mL whole blood processed.

  • The EasySep™ Direct HLA B Cell Isolation Kit can be used to isolate highly purified B cells directly from human whole blood, buffy coat, spleen, or lymph nodes by immunomagnetic negative selection. 
  • This kit targets non-B cells for removal with antibodies recognizing specific surface markers. Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep magnet.
  • Desired cells are simply collected into a new tube and can directly be used for downstream applications such as flow cytometry crossmatch.

Specifications

Cell Type B Cells
For Use With (Application) Cell Isolation
Quantity 100 mL whole blood
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.