Få mere at vide
Få mere at vide
Greiner Bio-One Easyload Pipette Tip Refill System, Sterile
Klik for at se tilgængelige muligheder
Volume (Metric):
200 μL
1000 μL
Pakningsstørrelse:
Pack of 2160
Pack of 5760
Beskrivelse
- Simple, time- and space-saving refill system for standard pipette tips.
- Autoclavable racks.
- Stackable racks.
- Color-coded boxes.
- Positioning guides make it easier to place refill units on the racks.
- Design of rack interior increases stability during autoclaving and when pipette tips are bein used with multi-channel pipettes.
- Graduated
- Sterile
- For single use only
- Polypropylene construction
- Free of heavy metal
Tekniske data
Tekniske data
| Volume (Metric) | 200 μL |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| Sterilization Method | Irradiation |
| Packaging | 960/Pk |
| Color Code | Yellow |
| Disposable | Single-use |
| For Use With (Equipment) | Suitable for single-channel Brand, Eppendorf, Finnpipette, Gilson, Socorex pipettes, multi-channel Brand, Gilson, Eppendorf, Socorex pipettes |
| Graduated | Yes |
| Product Line | EasyLoad |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion