missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One EasyLoad™ 200 Refill Universal Pipette Tips
Description
- Autoclavable EasyLoad™ racks
- Racks are stackable
- Color-coded boxes
- Design of rack's interior increases stability during autoclaving and when pipette tips are being used with multi-channel pipettes
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Sterility | Non-sterile |
| Color | Yellow |
| Autoclavable | Autoclavable |
| Format | Refill |
| For Use With (Equipment) | Socorex, Eppendorf, Gilson Pipetman |
| Graduated | Yes |
| Product Line | EasyLoad |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?