Learn More
E6130, MedChemExpress
Product Code. 16476837
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16476837
Brand: Medchem Express HY107456
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress E6130 is an orally active and highly selective CX3CR1 modulator, that may be effective for treatment of inflammatory bowel disease.
E6130 is an orally active and highly selective CX3CR1 modulator, that may be effective for treatment of inflammatory bowel disease.
Specifications
Chemical Name or Material E6130
CAS 1427058-33-0
Color White
Purity Grade Notes Research
Molecular Formula C28H37ClF3N3O3
Quantity 5 mg
Solubility Information DMSO : ≥ 250 mg/mL (449.59 mM)
SMILES C[C@@H](C1)[C@@](C(NC2CCN(CC3=CCCCC3)CC2)=O)(CC(O)=O)CN1CC4=C(C(F)(F)F)C=CC=C4Cl
Molecular Weight (g/mol) 556.06
Formula Weight 556.06
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation