Learn More
(E,E)-Farnesyl Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | trans, trans Farnesyl Bromide |
| CAS | 28290-41-7 |
| Molecular Formula | C15 H25 Br |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C15H25Br/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11H,5-6,8,10,12H2,1-4H3/b14-9+,15-11+ |
| SMILES | CC(=CCC\C(=C\CC\C(=C\CBr)\C)\C)C |
| IUPAC Name | (2E,6E)-1-bromo-3,7,11-trimethyldodeca-2,6,10-triene |
| Molecular Weight (g/mol) | 285.26 |
| Formula Weight | 284.114 |