Learn More
(E,E)-Farnesyl Bromide, TRC
Product Code. 30394119
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30394119 Supplier Toronto Research Chemicals Supplier No. TRCF1024401G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material trans, trans Farnesyl Bromide
CAS 28290-41-7
Molecular Formula C15 H25 Br
Quantity 1 g
InChI Formula InChI=1S/C15H25Br/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11H,5-6,8,10,12H2,1-4H3/b14-9+,15-11+
SMILES CC(=CCC\C(=C\CC\C(=C\CBr)\C)\C)C
IUPAC Name (2E,6E)-1-bromo-3,7,11-trimethyldodeca-2,6,10-triene
Molecular Weight (g/mol) 285.26
Formula Weight 284.114