Learn More
(E)-Dodec-2-enoic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (E)-Dodec-2-enoic Acid Ethyl Ester |
| CAS | 28290-90-6 |
| Molecular Formula | C14H26O2 |
| Quantity | 250 mg |
| Synonym | (2E)-2-Dodecenoic Acid Ethyl Ester, Ethyl (E)-2-Dodecenoate, Ethyl trans-2-Dodecenoate |
| InChI Formula | InChI=1S/C14H26O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h12-13H,3-11H2,1-2H3/b13-12+ |
| SMILES | C(/C=C/CCCCCCCCC)(OCC)=O |
| IUPAC Name | ethyl (E)-dodec-2-enoate |
| Molecular Weight (g/mol) | 226.36 |
| Formula Weight | 226.16 |