Learn More
(E)-Diethylstilbestrol D8 (ring-3,3',5,5'-diethyl-1,1,1',1'-D8) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30322464
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322464 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322464 Supplier Dr. Ehrenstorfer Supplier No. DREA12607020AL100
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material trans-Diethyl-1,1,1',1'-stilbestrol-3,3',5,5'-d8
CAS 91318-10-4
Solvent or Matrix Acetonitrile
Molecular Formula C18 2H8 H12 O2
Quantity 1 mL
Synonym Phen-2,6-d2-ol, 4,4'-[(1E)-1,2-di(ethyl-1,1-d2)-1,2-ethenediyl]bis-, Phen-2,6-d2-ol, 4,4'-[1,2-di(ethyl-1,1-d2)-1,2-ethenediyl]bis-, (E)- (9CI), 4,4'-[(1E)-1,2-Di(ethyl-1,1-d2)-1,2-ethenediyl]bis[phen-2,6-d2-ol], trans-Diethyl-1,1,1',1'-stilbestrol-3,3',5,5'-d8, (E)-Diethyl-1,1,1',1'-d4-stilbestrol-3,3',5,5'-d4, (E)-Diethylstilbestrol D8, (E)-α,α'-Diethylstilbene-4,4'-diol D8, Diethylstilbestrol (ring-3,3',5,5'-diethyl-1,1,1',1'-D8)
InChI Formula InChI=1S/C18H20O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,19-20H,3-4H2,1-2H3/b18-17+/i3D2,4D2,9D,10D,11D,12D
SMILES [2H]c1cc(cc([2H])c1O)\C(=C(\c2cc([2H])c(O)c([2H])c2)/C([2H])([2H])C)\C([2H])([2H])C
IUPAC Name 2,6-dideuterio-4-[(E)-2,2,5,5-tetradeuterio-4-(3,5-dideuterio-4-hydroxyphenyl)hex-3-en-3-yl]phenol
Molecular Weight (g/mol) 276.4
Formula Weight 276.1965 g/mol