Learn More
E-Ceftazidime, TRC
Product Code. 30136435
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136435 5 mg 5mg
30135715 10 mg 10mg
30008240 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136435 Supplier Toronto Research Chemicals Supplier No. TRCC2441255MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 97148-38-4
Molecular Formula C22H22N6O7S2
Molecular Weight (g/mol) 546.58
Synonym (6R,7R)-7-((E)-2-(2-Aminothiazol-4-yl)-2-(((2-carboxypropan-2-yl)oxy)imino)acetamido)-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Ceftazidime Impurity B
IUPAC Name (6R,7R)-7-[[(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-(2-carboxypropan-2-yloxyimino)acetyl]amino]-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CC(C)(O\N=C(\C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])C[n+]3ccccc3)/c4csc(N)n4)C(=O)O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C22H22N6O7S2/c1-22(2,20(33)34)35-26-13(12-10-37-21(23)24-12)16(29)25-14-17(30)28-15(19(31)32)11(9-36-18(14)28)8-27-6-4-3-5-7-27/h3-7,10,14,18H,8-9H2,1-2H3,(H4-,23,24,25,29,31,32,33,34)/b26-13+/t14-,18-/m1/s1
Formula Weight 546.0991
Chemical Name or Material E-Ceftazidime