Learn More
Vilber E-Box CX5 with UV transiluminator (EU plug)
Product Code. 31703657
Change view
Click to view available options
For Use With (Application):
Gel Documentation
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
31703657 Gel Documentation 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31703657 Supplier Vilber Supplier No. PF250200
  • ONLINE EXCLUSIVE PRICE
    10647.27 €
    8443.30 € / 1 sets
    Save 2203.97 €
    20% Off
Estimated Shipment: 31-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for precise imaging performance, it delivers accurate visualization and intuitive operation through a fast, user-friendly interface.
  • High-Quality Imaging: Scientific-grade camera made in Germany provides precise detection and high-resolution images for accurate gel analysis.
  • Exceptional Sensitivity: Detects very faint DNA, RNA, or protein bands, ensuring dependable performance with low-abundance samples.
  • Publication-Ready Results: Offers TIFF outputs that support scientific publication requirements while maintaining clarity and detail.
  • Clear Band Separation: High resolution and advanced imaging technology enable clear separation of closely spaced bands for reliable interpretation.
  • Large Touchscreen Display: Very large 12.6-inch touchscreen with Magster technology delivers fast, accurate, and intuitive navigation.
  • Built-in Industrial PC (Win10): Integrated industrial computer ensures rapid data processing and full control without external equipment.
  • Durable Construction: Steel and stainless-steel darkroom design provides long-lasting performance and easy cleaning.
  • Advanced Camera Features: Includes a 5-megapixel sensor expandable to 20 MP, passive cooling, USB3 connection, 16-bit imaging with 65,536 grey levels, and a motorized zoom lens.
  • Complete and Practical: Compact footprint fits standard lab benches and offers network connectivity, 3D Dynamic Scan, and BioVision software for imaging analysis.
  • Ease of Use: Stand-alone operation with one-click capture, auto-exposure, automatic illumination control, and UV shut-off after inactivity.
  • Time-Saving: Automatic functions and intuitive interface reduce manual steps and simplify experimental workflows.
  • Safe and Reliable: Automatic UV shut-off enhances safety and protects samples from unnecessary exposure.
  • Versatile Applications: Suitable for imaging DNA, RNA, and protein gels in research, teaching, and diagnostic laboratories.
Specifications
Filter Type Ethidium Bromide and Coomassie Blue
Pixel Size 5 MP
Includes EU Plug
For Use With (Application) Gel Documentation
Product Type Gel Documentation System
Display Type Touchscreen