Learn More
(E)-Acetylacrylic acid, TRC
Specifications
Specifications
| Chemical Name or Material | (E)-Acetylacrylic acid |
| CAS | 2833-28-5 |
| Molecular Formula | C5H6O3 |
| Quantity | 250 mg |
| Synonym | (2E)-4-Oxo-2-pentenoic Acid, (2E)-4-Oxopent-2-enoic Acid, (E)-3-Acetylpropenoic Acid, (E)-4-Oxopent-2-enoic Acid, (E)-4-Oxopent-2-enoic Acid, trans-3-Acetylacrylic Acid, trans-4-Oxo-2-pentenoic Acid, trans-beta-Acetylacrylic Acid |
| InChI Formula | InChI=1S/C5H6O3/c1-4(6)2-3-5(7)8/h2-3H,1H3,(H,7,8)/b3-2+ |
| SMILES | CC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-4-oxopent-2-enoic acid |
| Molecular Weight (g/mol) | 114.1 |
| Formula Weight | 114.0317 |