Learn More
3M™ E-A-R™ Classic™ SuperFit™ Refill Ear Plug Bottle
Product Code. 15789837
Change view
Click to view available options
:
Color: Yellow, Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15789837 Color: Yellow, Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15789837 Supplier 3M™ Supplier No. 7000089399
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ E-A-R™ Classic™ SuperFit™ Refill Ear Plug Bottle with a unique fitting ring to easily see when the earplug is correctly fitted.
  • Refill bottle with 500 3M™ E-A-R™ Classic™ SuperFit™ 33 Earplugs for 3M™ One-Touch earplug dispenser
  • Earplug with a orange fitting ring so its easy to when the earplug is correctly fitted
  • When the orange color is no longer visible, the best fit is achieved
  • Earplug dispenser serves as a reminder to wear hearing protection and make earplugs readily available in noisy areas for full time users and visitors alike
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow, Orange
Packaging Type Refill Bottle
Pairs per Package 500 Pairs/Bottle
Product Type Earplugs
Dielectric No