Learn More
(E)-[6]-Dehydroparadol, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
Description
(E)-[6]-Dehydroparadol, an oxidative metabolite of [6]-Shogaol (HY-14616), is a potent Nrf2 activator. (E)-[6]-Dehydroparadol can inhibit the growth and induce the apoptosis of human cancer cells.
Specifications
Specifications
| Chemical Name or Material | (E)-[6]-Dehydroparadol |
| CAS | 878006-06-5 |
| Color | Orange |
| Purity Grade Notes | Research |
| Molecular Formula | C17H24O3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 140 mg/mL (506.57 mM) |
| SMILES | CCCCCCCC(/C=C/C1=CC=C(O)C(OC)=C1)=O |
| Molecular Weight (g/mol) | 276.37 |
| Formula Weight | 276.37 |
| Show More |