missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (E)-3-Dimethylamino-1-(2-pyridyl)-2-propen-1-one, 98%

Product Code. 15447875
Click to view available options
Quantity:
5g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15447875

missing translation for 'mfr': Alfa Aesar™ H31714.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 66521-54-8
Molecular Formula C10H12N2O
MDL Number MFCD00115039
Quantity 5g
Synonym e-3-dimethylamino-1-pyridin-2-yl prop-2-en-1-one, 3-dimethylamino-1-pyridin-2-yl prop-2-en-1-one, 2e-3-dimethylamino-1-pyridin-2-yl prop-2-en-1-one, 3-dimethylamino-1-pyridin-2-yl-propenone, 2-3-dimethylamino acryloyl pyridine, 2e-3-dimethylamino-1-2-pyridyl-2-propen-1-one, 3-dimethylamino-1-2-pyridyl-2-propen-1-one, 1-2-pyridyl-3-dimethylamino-2-propen-1-one, pubchem23485, 2-propen-1-one,3-dimethylamino-1-2-pyridinyl
InChI Key BWERGHWJEBQNQV-SOFGYWHQSA-N
SMILES CN(C)C=CC(=O)C1=CC=CC=N1
IUPAC Name (E)-3-(dimethylamino)-1-pyridin-2-ylprop-2-en-1-one
Molecular Weight (g/mol) 176.219
PubChem CID 5369159
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material (E)-3-Dimethylamino-1-(2-pyridyl)-2-propen-1-one
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.