Learn More
(E)-3-Bromoacrylic Acid, TRC
Product Code. 30335404
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335404 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30335404 Supplier Toronto Research Chemicals Supplier No. TRCB679400500MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (E)-3-Bromoacrylic Acid
CAS 6213-89-4
Molecular Formula C3H3O2Br
Quantity 500 mg
InChI Formula InChI=1S/C3H3BrO2/c4-2-1-3(5)6/h1-2H,(H,5,6)/b2-1+
SMILES OC(=O)\C=C\Br
IUPAC Name (E)-3-bromoprop-2-enoic acid
Molecular Weight (g/mol) 150.96
Formula Weight 149.9316