Learn More
(E)-2-Octadecenoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (E)-2-Octadecenoic Acid |
| CAS | 2825-79-8 |
| Molecular Formula | C18 H34 O2 |
| Quantity | 5 mg |
| Synonym | trans-2-Octadecenoic Acid, (2E)-2-Octadecenoic Acid |
| InChI Formula | InChI=1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h16-17H,2-15H2,1H3,(H,19,20)/b17-16+ |
| SMILES | CCCCCCCCCCCCCCC\C=C\C(=O)O |
| IUPAC Name | (E)-octadec-2-enoic acid |
| Molecular Weight (g/mol) | 282.461 |
| Formula Weight | 282.256 |