Learn More
(E)-1-Ethoxy-1-penten-3-one (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | (E)-1-Ethoxy-1-penten-3-one |
| CAS | 79317-83-2 |
| Molecular Formula | C7H12O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C7H12O2/c1-3-7(8)5-6-9-4-2/h5-6H,3-4H2,1-2H3/b6-5+ |
| SMILES | CCO\C=C\C(=O)CC |
| IUPAC Name | (E)-1-ethoxypent-1-en-3-one |
| Molecular Weight (g/mol) | 128.17 |
| Formula Weight | 128.0837 |