Learn More
Dysferlin Antibody (CL10888) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07994AF700
Product Code. 30296744
  • 737.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Dysferlin Monoclonal antibody specifically detects Dysferlin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dysferlin
Monoclonal
Alexa Fluor 700
dysferlin, dysferlin, limb girdle muscular dystrophy 2B (autosomal recessive), dystrophy-associated fer-1-like 1, FER1L1Dystrophy-associated fer-1-like protein, Fer-1-like protein 1, FLJ00175, FLJ90168, LGMD2B
This antibody was developed using a recombinant protein derived from O75923, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers
8291
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL10888
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified