missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYRK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540341
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GDHTQHFLDG GEMKVEQLFQ EFGNRKSNTI QSDGISDSEK CSPTVSQGKS Sequence homology: Human: 100%; Mouse: 86%
DYRK3 Polyclonal antibody specifically detects DYRK3 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
DYRK3 | |
Polyclonal | |
Unconjugated | |
DYRK3 | |
BC006704; dual specificity tyrosine phosphorylation regulated kinase 3; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual specificity tyrosine-phosphorylation-regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5; dyrk3; dyrk3 {ECO:0000312; DYRK5; EMBL:AAH97474.1}; hYAK3-2; KSR; protein kinase Dyrk3; RED; REDK; regulatory erythroid kinase; RP11-343H5.2; RSU2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8444 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43781 | |
DYRK3 | |
Synthetic peptide directed towards the N-terminal of human DYRK3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |