missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynamin 2 Monoclonal Antibody (3F5C7)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA548443

Product Code. 17955861

  • 512.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Dynamin 2 Monoclonal Antibody for Western Blot, ELISA

Dynamins represent one of the subfamilies of GTP-binding proteins. These proteins share considerable sequence similarity over the N-terminal portion of the molecule, which contains the GTPase domain. Dynamins are associated with microtubules. They have been implicated in cell processes such as endocytosis and cell motility, and in alterations of the membrane that accompany certain activities such as bone resorption by osteoclasts. Dynamins bind many proteins that bind actin and other cytoskeletal proteins. Dynamins can also self-assemble, a process that stimulates GTPase activity. Five alternatively spliced transcripts encoding different proteins have been described. Additional alternatively spliced transcripts may exist, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dynamin 2
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P50570
DNM2
Purified recombinant fragment of human Dynamin-2 (AA: 520-744) expressed in E. Coli.
100 μg
Primary
Human
Antibody
IgG2a
ELISA, Western Blot, Immunocytochemistry, Immunohistochemistry, Western Blot
3F5C7
Unconjugated
DNM2
b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; Dnm2; DYIIAAB; Dyn2; dynamin 2; dynamin II; Dynamin UDNM; Dynamin-2; DYNII; LCCS5; Udnm
Mouse
Protein G
RUO
1785
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.