missing translation for 'onlineSavingsMsg'
Learn More

Dynamin 2 Antibody (3F5F3) - BSA Free, Novus Biologicals™

Product Code. 18272954 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18272954 0.1 mL 0.1mL
18673048 0.025 mL 0.03mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18272954 Supplier Novus Biologicals Supplier No. NBP261757

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dynamin 2 Monoclonal specifically detects Dynamin 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynamin 2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 3F5F3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII
Gene Symbols DNM2
Host Species Mouse
Immunogen Purified recombinant fragment of human Dynamin-2 (AA: 520-744) expressed in E. Coli.
Molecular Weight of Antigen 98 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1785
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.