Learn More
Invitrogen™ Dynabeads™ Untouched™ Human CD4 T Cells Kit
Use this kit to isolate pure and viable untouched CD4+ T-cells from PBMC by negative isolation. The kit depletes CD8+ T cells, B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes and erythrocytes.
309.00 € - 1042.00 €
Specifications
Cell Type | T Cells (CD4) |
---|---|
Product Type | Cell Isolation Kit |
Final Product Type | Cells |
Isolation Technology | Negative Isolation |
Product Line | DYNAL™, Dynabeads™, Untouched™ |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10480065
|
Invitrogen™
11352D |
2 mL |
309.00 €
2mL |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10449053
|
Invitrogen™
11346D |
10 mL |
1042.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
The kit depletes CD8+ T cells, B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes and erythrocytes. The negatively isolated human CD4+ T-cells are left in sample and have not been in contact with Dynabeads. The bead and antibody-free CD4+ T-cells are truly untouched, and in perfect shape for any functional assay and application.
- High purity, recovery and viability of human untouched CD4+ T cells
- Easy to use and to scale up
- No columns required
- Not high throughput compatible
A negative isolation strategy isolates pure and truly untouched cells. When maximum cell purity is critical, a positive cell isolation strategy is recommended.
Specifications
T Cells (CD4) | |
Cells | |
DYNAL™, Dynabeads™, Untouched™ | |
Room Temperature |
Cell Isolation Kit | |
Negative Isolation | |
Research Grade | |
Human |
For Research Use Only. Not for use in diagnostic procedures.