Learn More
Applied Biosystems™ Dynabeads™ MPC™-S (Magnetic Particle Concentrator)
Our Magnetic Particle Concentrators (MPC™) enable optimal binding of Dynabeads™ magnetic beads with their targets in a variety of starting samples.
Brand: Applied Biosystems™ A13346
Description
The MPC™-S was designed specifically for microbiology applications; it has been validated with select Dynabeads™ for enrichment of E. coli O157:H7, Salmonella, non-O157 STEC, Cryptosporidium, and Giardia. The MPC™-S can be used with both food enrichment samples types as well as water samples. It holds up to six microcentrifuge tubes and has an optimal working volume range of 20 µl–2 ml.
Note that this magnet is for microbiology applications only, not for eukaryotic cell separations. For use in food and environmental testing only. Not for any animal or human therapeutic or diagnostic use.
Specifications
| Store at room temperature. | |
| Dynabeads™ MPC™, Dynabeads™ Sample Mixers | |
| Dynabeads™ | |
| Bacterial Culture |
| 6 Microtubes | |
| Not High-throughput Compatible (Manual) | |
| 1 to 6 microfuge tubes, Up to 2 mL/tube | |
| Room Temperature |