Learn More
Thermo Scientific™ DyLight™ 680-4xPEG Amine-Reactive Dye and Kits
Shop All Thermo Scientific ProductsDescription
Fluorescently label antibodies and other proteins with Thermo Scientific™ DyLight 680-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 680 Dye.
Additional Information and Online Ordering
Highlights:
High fluorescence intensity – significantly brighter fluorescence than Alexa Fluor™ 680
PEGylated – improves solubility in aqueous solution, aids in cell permeability and improves tissue retention in in vivo applications
Convenient kit sizes – standard and microscale kit sizes to match your experimental needs
Recommended for:
- Fluorescence microscopy
- In vivo or ex vivo imaging
- Cell-based assays
- Flow cytometry/fluorescence-activated cell sorting (FACS)
Specifications
Specifications
| Formulation | NHS ester activated proprietary near-infrared fluorescent dye |
| Quantity | 5 x 65 μg |
| Product Type | DyLight 680-4xPEG NHS Ester |
| Sufficient For | 5 uses to label a total of 2mg of IgG using typical conditions |
| Product Line | DyLight |