missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dye Primer/Matrix Standards [5FAM™, JOE™, TAMRA™, and ROX™ dyes]

Product Code. 15889706 Shop All Applied Biosystems Products
This item is not returnable. View return policy

Product Code. 15889706

Brand: Applied Biosystems™ 401114

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Contains dye labeled standards used to generate multicomponent matrix. 1 tube of each dye

The standard can be used for Dye Primer cycle Sequencing and Dye Primer Sequenase Sequencing and on the 310, 3100/3100-Avant™ and 3130/XL Analyzers.

This Dye Primer Matrix Standards are used to generate the multicomponent matrix required when analyzing 5-FAM™, JOE™, TAMRA™ and ROX™-labeled DNA fragments.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains dye labeled standards used to generate multicomponent matrix. 1 tube of each dye. Store the kit at 2 - 8°C. Do not freeze.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Label or Dye FAM, JOE, ROX, TAMRA™
Product Type DNA Seqeuncing Size Standard
Quantity 1 Ea.
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.