Learn More
DY131, MedChemExpress
Product Code. 16452657
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452657 50 mg 50mg
16436568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16452657 Supplier Medchem Express Supplier No. HY15483
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress DY131 (GSK 9089) is a potent and selective ERRγ and ERRβ agonist. DY131displays inactive against ERRα, ERα and ERβ. DY131 also inhibits Smo signaling.
DY131 (GSK 9089) is a potent and selective ERRγ and ERRβ agonist. DY131displays inactive against ERRα, ERα and ERβ. DY131 also inhibits Smo signaling.
Specifications
Chemical Name or Material DY131
CAS 95167-41-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H21N3O2
Quantity 50 mg
Synonym GSK 9089
Solubility Information DMSO : 50 mg/mL (160.58 mM; Need ultrasonic)
SMILES O=C(N/N=C/C1=CC=C(N(CC)CC)C=C1)C2=CC=C(O)C=C2
Molecular Weight (g/mol) 311.38
Formula Weight 311.38
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death