missing translation for 'onlineSavingsMsg'
Learn More

DUX4 Antibody (P4H2) - Azide and BSA Free, R&D Systems™

Product Code. 18240057 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18240057 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18240057 Supplier Novus Biologicals Supplier No. NBP280703
  • ONLINE EXCLUSIVE PRICE
    561.95 €
    477.75 € / 0.1mL
    Save 84.20 €
    14% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

DUX4 Monoclonal specifically detects DUX4 in Human samples. It is validated for ELISA/Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Immunohistochemistry (Frozen)/Immunohistochemistry (Paraffin)/Simple Western/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUX4
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Monoclonal
Clone P4H2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 0.5-2.0ug/ml, Simple Western : 1:10, ELISA reported in scientific literature (PMID 21529284), Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence : 1:100-1:200, Immunohistochemistry-Paraffin : 1:100-1:200, Immunohistochemistry-Frozen : 1:100-1:200
Formulation PBS
Gene Alias double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10
Gene Symbols DUX4
Host Species Mouse
Immunogen Synthetic peptide corresponding to the C-terminus of human DUX4. [Swiss-Prot: Q9UBX2]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 100288687
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.