missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Duvakitug Humanized Recombinant Human Monoclonal Antibody
Description
Compatible with Direct ELISA.
Duvakitug inhibits the enzyme responsible for the synthesis of pro-inflammatory cytokines, thereby reducing inflammation.
Specifications
Specifications
| Antigen | Duvakitug Humanized |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | SAR-447189; TEV'574; TEV-48574 |
| Host Species | Human |
| Immunogen | Recombinant Human TNFSF15/TL1A. |
| Purification Method | Protein A/G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?